Learn More
Poltex Locking N95 Mask Dispenser
SDP
Catalog No. 22000462
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-462 Wall Mount
22-000-463 VHB Tape
22-000-464 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-462 Supplier Poltex Supplier No. N95MSKLCKW
Only null left
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Holds and dispenses one box of N95 Masks.
  • Made with extremely durable, break resistant 3/16 in. thick PETG
  • Choose from keyhole slots for wall mounting, or powerful magnets.
  • Locking, sliding lid keeps dust out
  • Hardware not included for wall mount unit (-W). Customer must provide their own screw/anchor for their wall type.
Specifications
Product Type Locking N95 Dispenser
Material PETG
For Use With (Application) Healthcare
For Use With (Equipment) N95 masks
Mounting Wall Mount