Learn More
Med Vet International Locking Plug for TPLO Locking Screw, J0438P
Catalog No. 5000582349
Encompass_Preferred
Used to protect locking holes while contouring plate
Catalog No. 50-005-82349 Supplier Med Vet International Supplier No. JOR438P
Used to protect locking holes while contouring plate