Learn More
Electron Microscopy Sciences Locking Procedure 6 Drawer Cart with Blue Drawers
SDP
Catalog No. 501927319
Encompass_Preferred
White Polyethylene Locking 6 Drawer Procedure Cart has six 24 inch wide lockable full extension drawers and heavy duty industrial casters with brakes. The top is a 3 inch deep tray with a built in handle for easy pushing or pulling. Cart features four 3 inch deep drawers and two 7 inch deep drawers. This chemically resistant cart features sturdy polyethylene, ABS and PVC construction.
Color: Blue Drawers
Catalog No. 50-192-7319 Supplier Electron Microscopy Sciences Supplier No. 6438681
White Polyethylene Locking 6 Drawer Procedure Cart has six 24 inch wide lockable full extension drawers and heavy duty industrial casters with brakes. The top is a 3 inch deep tray with a built in handle for easy pushing or pulling. Cart features four 3 inch deep drawers and two 7 inch deep drawers. This chemically resistant cart features sturdy polyethylene, ABS and PVC construction.
Color: Blue Drawers