Learn More
Med Vet International Locking Screw, J1220L, 2.4mm x 30mm, Each
Catalog No. 5000581180
Encompass_Preferred
Jorvet 2.4 twin start locking screws (J1220 series) are fully compatible with the industry standard. The 2.4 locking plates (J1220 series) are available in 4 to 18 hole sizes.2.4mm Locking Screw 2.4 head 30mm long
Catalog No. 50-005-81180 Supplier Med Vet International Supplier No. JOR1220L
Jorvet 2.4 twin start locking screws (J1220 series) are fully compatible with the industry standard. The 2.4 locking plates (J1220 series) are available in 4 to 18 hole sizes.2.4mm Locking Screw 2.4 head 30mm long