Learn More
Med Vet International Locking Screw, J1220O, 2.4mm x 36mm, Each
Catalog No. 5000581112
Encompass_Preferred
Jorvet 2.4 twin start locking screws (J1220 series) are fully compatible with the industry standard. The 2.4 locking plates (J1220 series) are available in 4 to 18 hole sizes.2.4mm Locking Screw 2.4 head 36mm long
Catalog No. 50-005-81112 Supplier Med Vet International Supplier No. JOR1220O
Jorvet 2.4 twin start locking screws (J1220 series) are fully compatible with the industry standard. The 2.4 locking plates (J1220 series) are available in 4 to 18 hole sizes.2.4mm Locking Screw 2.4 head 36mm long