Learn More
Post Medical Locking-Top Sharps Containers
Catalog No. PMSL150
Change view
Click to view available options
Capacity (English):
1.5 qt.
2 qt.
1.5 gal.
2.5 gal.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
PMSL150 1.5 qt.
PMSL200 2 qt.
PM1101LPR 2.5 gal.
PM2201LPR 1.5 gal.
PM1101LPBW6 2.5 gal.
PM2201LPBW8 1.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PMSL150 Supplier Post Medical Supplier No. SL150
Add to Cart
Add to Cart
Safe, one-handed sharps disposal
For use in patient rooms and all hospital departments including laboratory.
  • Thick-walled, puncture-resistant sharps containment systems provide safe, convenient and affordable site of origin sharps disposal
  • Vertical drop disposal minimizes excessive handling; reduces risks associated with horizontal “mail-box” type disposal
  • Once self-contained screw cap is secured and locked, leakage of fluids during handling, transport and disposal is virtually eliminated
  • 1.0, 1.5 and 2.5 gal. Containers
  • Can be used freestanding or with metal stands or cabinets
  • Bulk 3.5 and 6 gal. Containers
  • Designed for freestanding use only
Compliance
Meets OSHA requirements.
Specifications
Capacity (Metric) 1.4 L
Closure Type Leaktight Locking Top, Snap-Tite Lid, Safe-T-Ring
Capacity (English) 1.5 qt.
For Use With (Application) For use in patient rooms and all hospital departments including laboratory.