Learn More
Marketlab, Inc. LockMailer Microscope Slide Jar
Catalog No. 22505482
Change view
Click to view available options
Slide Capacity:
4
Catalog No. Slide Capacity
22-505-482 4
1 options
Catalog No. 22-505-482 Supplier Marketlab, Inc. Supplier No. 0970
Add to Cart
Add to Cart
Locking Microscope Slide Jar for Mailing, Staining, and Storage.
  • Can be used with and without locking feature
  • Polypropylene vial holds four 75 x 25 mm slides
  • Caps are leakproof HDPE
  • Optional Capinserts color code specimens to meet your needs
Specifications
Type Slide Mailer
Slide Capacity 4