Learn More
Research Products International Corp Lockmailer Tube, 4 Slide Capacity, 100/pk
SDP
Catalog No. 501978324
Encompass
LockMailer Tube is a multipurpose container for mailing, staining or storing microscope slides. Designed with a unique tamper evident screw cap that ensures sample integrity during all phases of storage and transport.
Free standing tube holds up to 4 standard 3 x 1" slides vertically with internal volume of 12ml. Slotted inner channels keep slides separate. Tubes are made of polypropylene and high density polyethylene cap.
Catalog No. 50-197-8324 Supplier Research Products International Corp Supplier No. 144580
LockMailer Tube is a multipurpose container for mailing, staining or storing microscope slides. Designed with a unique tamper evident screw cap that ensures sample integrity during all phases of storage and transport.
Free standing tube holds up to 4 standard 3 x 1" slides vertically with internal volume of 12ml. Slotted inner channels keep slides separate. Tubes are made of polypropylene and high density polyethylene cap.