Learn More
Grainger Lockout Padlock KA Silver 1-1/2 H PK12
Catalog No. 502764479
Encompass_Preferred
The American Lock A1106RED solid brass padlock has a 1-1/16" wide red plastic coated shackle that provides visibility and corrosion resistance. It features a 1-1/4" hardened steel shackle for strength and security. The padlock body is made from solid brass for durability. This padlock has a double locking mechanism with a 5-pin tumbler cylinder to provide pick and drill resistance. It can be keyed differently or keyed alike for convenience. The American Lock A1106RED padlock is ideal for lockout/tagout applications.
Catalog No. 50-276-4479 Supplier Grainger Supplier No. 16P023
The American Lock A1106RED solid brass padlock has a 1-1/16" wide red plastic coated shackle that provides visibility and corrosion resistance. It features a 1-1/4" hardened steel shackle for strength and security. The padlock body is made from solid brass for durability. This padlock has a double locking mechanism with a 5-pin tumbler cylinder to provide pick and drill resistance. It can be keyed differently or keyed alike for convenience. The American Lock A1106RED padlock is ideal for lockout/tagout applications.