Learn More
Brady™ Lockout Point Label Polyester with Overlaminate
Catalog No. 19129949 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Polyester
Catalog No. Material
19-129-949 Polyester
1 options
Catalog No. 19-129-949 Supplier Brady™ Supplier No. 151254
Add to Cart
Add to Cart
Get the identification you need to clearly identify lockout points on your equipment.
  • General lockout point identification labels can be used to identify a lockout point without needing to worry about point numbering or type of hazard
  • OSHA compliant
  • B-302 is an indoor/outdoor high-performance pressure-sensitive material, and is surface printed polyester with a polyester overlaminate
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
Specifications
Product Type Lockout Point Label
Color Red/White
Material Polyester
Height (English) 1.5 in.
Quantity 25/Pk.
Temperature Range (English) −40° to +110°C
Width (English) 3 in.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.