Learn More
Brady™ Lockout Procedure Station with Binder
Catalog No. 19103197 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-197 Red
1 options
Catalog No. 19-103-197 Supplier Brady™ Supplier No. 99289
Add to Cart
Add to Cart
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Lockout Station is perfect for ensuring that all of lockout procedures are readily available to employees. The procedure station's silk-screened backboard is made from durable polystyrene and includes pre-drilled holes and hardware for wall mounting.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Procedure Station
Color Red
Material Polystyrene
Includes Backboard with white PVC coated steel binder rack and hardware and a Lockout Procedure Binder (LOSB1). (Procedure Forms sold separately)
Quantity 1 Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.