Learn More
Waters Corp Waters LockSpray Probe Tip Assembly, Replacement, Mass Spectrometry
Catalog No. 50793328 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement LockSpray Probe Tip Assembly designed for use with specific Waters mass spectrometry instruments. It ensures proper functionality and maintenance of your analytical equipment.
  • Replacement probe tip assembly
  • Utilized on LCT Premier, Qtof Premier, Synapt Series, Xevo Qtof Series, Vion instruments
  • Part of spare parts kits
Catalog No. 50-793-328 Supplier Waters Corp Supplier No. M955615BC2
This is a replacement LockSpray Probe Tip Assembly designed for use with specific Waters mass spectrometry instruments. It ensures proper functionality and maintenance of your analytical equipment.
  • Replacement probe tip assembly
  • Utilized on LCT Premier, Qtof Premier, Synapt Series, Xevo Qtof Series, Vion instruments
  • Part of spare parts kits