Learn More
Waters Corp Waters LockSpray Reference Probe Assembly, Mass Spectrometry, Replacement
Catalog No. 50793019 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement LockSpray Reference Probe Assembly utilized on Synapt, Xevo, and Vion LockSpray sources for mass spectrometry systems.
  • Designed for mass spectrometry systems.
  • Compatible with Synapt, Xevo, and Vion LockSpray sources.
  • Replacement part, for use as required.
  • Includes one probe assembly per package.
Catalog No. 50-793-019 Supplier Waters Corp Supplier No. 700004588
This is a replacement LockSpray Reference Probe Assembly utilized on Synapt, Xevo, and Vion LockSpray sources for mass spectrometry systems.
  • Designed for mass spectrometry systems.
  • Compatible with Synapt, Xevo, and Vion LockSpray sources.
  • Replacement part, for use as required.
  • Includes one probe assembly per package.