Learn More
RS Hughes Inc LOCKTITE 495 1OZ SUPERGLUE LIQ
SDP
Catalog No. NC9011323
Encompass_Preferred
Loctite Super Bonder 495 Cyanoacrylate Adhesive - Clear Liquid 1 oz Bottle - Shear Strength 2750 psi, Tensile Strength 1740 to 3625 psi
Catalog No. NC9011323 Supplier RS Hughes Inc Supplier No. 07934049550
Loctite Super Bonder 495 Cyanoacrylate Adhesive - Clear Liquid 1 oz Bottle - Shear Strength 2750 psi, Tensile Strength 1740 to 3625 psi