Learn More
Grainger Malin Company Lockwire, Stainless Steel, 0.02 in. Diameter, 931 ft.
Catalog No. 502766875
Encompass_Preferred
This stainless steel lockwire is a reliable tool designed to secure bolts and fasteners in high-stress environments. Functioning as a positive locking device, it prevents fasteners from loosening or detaching under vibration or extreme conditions. The wire is constructed from stainless steel type 302/304, meeting ASTM A 580 standards, and provides effective corrosion resistance for demanding applications including aerospace and industrial machinery.
  • Features a nominal diameter of 0.02 in. and a total length of 931 ft.
  • Constructed from corrosion-resistant stainless steel type 302/304 per ASTM A 580.
  • Includes a tensile strength of 75,000 psi.
  • Supplied in a recyclable canister for convenient storage and usage.
  • Easy to shape and form for versatile installation requirements.
  • Colour-coded for easy identification.
Catalog No. 50-276-6875 Supplier Grainger Supplier No. 16Y047
This stainless steel lockwire is a reliable tool designed to secure bolts and fasteners in high-stress environments. Functioning as a positive locking device, it prevents fasteners from loosening or detaching under vibration or extreme conditions. The wire is constructed from stainless steel type 302/304, meeting ASTM A 580 standards, and provides effective corrosion resistance for demanding applications including aerospace and industrial machinery.
  • Features a nominal diameter of 0.02 in. and a total length of 931 ft.
  • Constructed from corrosion-resistant stainless steel type 302/304 per ASTM A 580.
  • Includes a tensile strength of 75,000 psi.
  • Supplied in a recyclable canister for convenient storage and usage.
  • Easy to shape and form for versatile installation requirements.
  • Colour-coded for easy identification.