Learn More
Grainger Malin Company Lockwire, Stainless Steel, 0.032 in. Diameter, 364 ft. Length, 1 lb. Canister
Catalog No. 502766823
Encompass_Preferred
This stainless steel safety wire is a positive locking device designed to ensure that fasteners remain tight and secure. It is specifically engineered to prevent fasteners from loosening or falling out due to vibration and operational stress. The wire is supplied in a durable, high density polyethylene canister that keeps it free of grease and corrosive materials while allowing for easy identification through color-coding.
  • Meets NASM/MS20995C and ASTM A580 specifications
  • Constructed from type 302/304 stainless steel
  • Annealed to spring temper
  • Tensile strength of 75,000 psi
  • Suitable for securing bolts on aircraft, engines, race cars, and industrial machinery
  • Recyclable, non-degrading canister features
  • Blue canister color-coding indicates stainless steel material
Catalog No. 50-276-6823 Supplier Grainger Supplier No. 16Y049
This stainless steel safety wire is a positive locking device designed to ensure that fasteners remain tight and secure. It is specifically engineered to prevent fasteners from loosening or falling out due to vibration and operational stress. The wire is supplied in a durable, high density polyethylene canister that keeps it free of grease and corrosive materials while allowing for easy identification through color-coding.
  • Meets NASM/MS20995C and ASTM A580 specifications
  • Constructed from type 302/304 stainless steel
  • Annealed to spring temper
  • Tensile strength of 75,000 psi
  • Suitable for securing bolts on aircraft, engines, race cars, and industrial machinery
  • Recyclable, non-degrading canister features
  • Blue canister color-coding indicates stainless steel material