Learn More
Grainger Lockwire Canister 0.041 Dia 221 ft.
Catalog No. 502766874
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a lock effect when installed in a crisscross pattern to secure bolts, screws, nuts and other threaded assemblies. It conforms to MIL-W-83420 and MS20995 standards. The lockwire is made in the USA.
Catalog No. 50-276-6874 Supplier Grainger Supplier No. 16Y050
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a lock effect when installed in a crisscross pattern to secure bolts, screws, nuts and other threaded assemblies. It conforms to MIL-W-83420 and MS20995 standards. The lockwire is made in the USA.