Learn More
Grainger MALIN COMPANY Lockwire, Inconel, 0.041 in. Diameter, 207.12 ft. Length, 1 ea.
Catalog No. 502766906
Encompass_Preferred
This safety lockwire is designed to secure fasteners against loosening in high-vibration and high-movement applications. It is threaded through a fastener, twisted tightly, and anchored to a secondary point to ensure structural integrity and stability.
  • Inconel material construction
  • 0.041 in. diameter wire
  • 207.12 ft. total length
  • Meets military specification MS20995N41
  • 80,000 to 105,000 PSI tensile strength
  • Supplied in a canister package
Catalog No. 50-276-6906 Supplier Grainger Supplier No. 16Y064
This safety lockwire is designed to secure fasteners against loosening in high-vibration and high-movement applications. It is threaded through a fastener, twisted tightly, and anchored to a secondary point to ensure structural integrity and stability.
  • Inconel material construction
  • 0.041 in. diameter wire
  • 207.12 ft. total length
  • Meets military specification MS20995N41
  • 80,000 to 105,000 PSI tensile strength
  • Supplied in a canister package