Learn More
Grainger Lockwire Spool 0.02 Dia 4655 ft.
Catalog No. 502765521
Encompass_Preferred
The Malin Company Lockwire is made of stainless steel and is 0.032 inches in diameter. It comes in a spool with a length of 100 feet. Lockwire is used to prevent loosening of assemblies because of vibration and rotation. It provides maximum locking security. This lockwire conforms to MIL-W-17337 standards. It can withstand high temperatures up to 1200°F. The stainless steel composition resists corrosion. This is an aviation-grade lockwire ideal for critical assemblies.
Catalog No. 50-276-5521 Supplier Grainger Supplier No. 16Y052
The Malin Company Lockwire is made of stainless steel and is 0.032 inches in diameter. It comes in a spool with a length of 100 feet. Lockwire is used to prevent loosening of assemblies because of vibration and rotation. It provides maximum locking security. This lockwire conforms to MIL-W-17337 standards. It can withstand high temperatures up to 1200°F. The stainless steel composition resists corrosion. This is an aviation-grade lockwire ideal for critical assemblies.