Learn More
Grainger Lockwire Spool 0.032 Dia 1820 ft.
Catalog No. 502767765
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a positive locking device when installed properly in bolts, nuts, turnbuckles etc. It conforms to MIL-W-83420 and MS20995 standards. The lockwire works by exerting a constant tension against the threaded fastener.
Catalog No. 50-276-7765 Supplier Grainger Supplier No. 16Y054
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a positive locking device when installed properly in bolts, nuts, turnbuckles etc. It conforms to MIL-W-83420 and MS20995 standards. The lockwire works by exerting a constant tension against the threaded fastener.