Learn More
Grainger Lockwire Spool 0.041 Dia 51.78ft
Catalog No. 502767954
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and is made in the USA. The lockwire creates an inexpensive locking device when installed in a crisscross pattern through bolt heads or nuts after torquing. It features excellent corrosion resistance and has aerospace, military, industrial, and motorsports applications. The lockwire meets QQ-W-406 and AMS-W-6845 specifications.
Catalog No. 50-276-7954 Supplier Grainger Supplier No. 16Y060
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and is made in the USA. The lockwire creates an inexpensive locking device when installed in a crisscross pattern through bolt heads or nuts after torquing. It features excellent corrosion resistance and has aerospace, military, industrial, and motorsports applications. The lockwire meets QQ-W-406 and AMS-W-6845 specifications.