Learn More
Grainger Lockwire Spool 0.041 Dia 55.25 ft.
Catalog No. 502766822
Encompass_Preferred
The Malin Company Lockwire is a 0.032" diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225' roll and is made in the USA. The lockwire creates an inexpensive locking device when installed in a crisscross pattern through the bolt heads or nuts of an assembly. It features excellent corrosion resistance and has aerospace grade tensile strength. This lockwire securely locks threaded fasteners in critical applications.
Catalog No. 50-276-6822 Supplier Grainger Supplier No. 16Y045
The Malin Company Lockwire is a 0.032" diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225' roll and is made in the USA. The lockwire creates an inexpensive locking device when installed in a crisscross pattern through the bolt heads or nuts of an assembly. It features excellent corrosion resistance and has aerospace grade tensile strength. This lockwire securely locks threaded fasteners in critical applications.