Learn More
Grainger Lockwire Spool 0.051 Dia 31.948 ft
Catalog No. 502765552
Encompass_Preferred
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a positive locking device when installed properly in bolts, nuts, turnbuckles and other threaded assemblies. It conforms to MIL-W-83420 specifications. The lockwire is made in the USA.
Catalog No. 50-276-5552 Supplier Grainger Supplier No. 16Y068
The Malin Company Lockwire is a 0.032 inch diameter stainless steel lockwire used to prevent loosening of threaded fasteners. It comes in a 225 foot roll and features a tensile strength of 300,000 psi. The lockwire creates a positive locking device when installed properly in bolts, nuts, turnbuckles and other threaded assemblies. It conforms to MIL-W-83420 specifications. The lockwire is made in the USA.