Learn More
Electron Microscopy Sciences Loctite 349™ Impruv 50 ML
SDP
Catalog No. 50949058
Encompass_Preferred
For bond glass/metal, appearance: clear/straw, cure type: UV, viscosity: 9,500cP, shore hardness: D70, temperature range: -65 deg F to 266 deg F.
Catalog No. 50-949-058 Supplier Electron Microscopy Sciences Supplier No. 7258201
For bond glass/metal, appearance: clear/straw, cure type: UV, viscosity: 9,500cP, shore hardness: D70, temperature range: -65 deg F to 266 deg F.