Learn More
RS Hughes Inc LOCTITE 3926 FLUORSC 25ML 10CS
SDP
Catalog No. NC2373505
Encompass_Preferred
Loctite 3926 fluorescent one-part acrylic adhesive is compatible with glass, metal, pvc and thermoplastic materials. Delivers great performance with a shear strength of 1330 psi and tensile strength of 2740 psi. Minimum to maximum operating temperatures are -65 F to +300 F. Comes in a 25 ml Syringe.Perfect for bonding applications. This acrylic adhesive is packaged 25 ml per inner, 10 per case. The IDH number for this item is 434108.
Catalog No. NC2373505 Supplier RS Hughes Inc Supplier No. 07934036492
Loctite 3926 fluorescent one-part acrylic adhesive is compatible with glass, metal, pvc and thermoplastic materials. Delivers great performance with a shear strength of 1330 psi and tensile strength of 2740 psi. Minimum to maximum operating temperatures are -65 F to +300 F. Comes in a 25 ml Syringe.Perfect for bonding applications. This acrylic adhesive is packaged 25 ml per inner, 10 per case. The IDH number for this item is 434108.