Learn More
Electron Microscopy Sciences Loctite 409™ Super Bonder Tube 3 GR
SDP
Catalog No. 50949015
Encompass_Preferred
General-purpose gel adhesive, clear, gap fills 0.010"; surface insensitive Ethyl, tensile shear strength 3,200 psi, and temperature range -65 deg F to 180 deg F. Cure speed: fixture - 75 seconds, full - 24 hours.
Catalog No. 50-949-015 Supplier Electron Microscopy Sciences Supplier No. 7257009
General-purpose gel adhesive, clear, gap fills 0.010"; surface insensitive Ethyl, tensile shear strength 3,200 psi, and temperature range -65 deg F to 180 deg F. Cure speed: fixture - 75 seconds, full - 24 hours.