Learn More
XPEDX LOCTITE 454
Catalog No. NC9613061
Encompass_Preferred
This item has a minimum qty of 41 per supplier requirements.
A general-purpose, no-run clear gel. Can be applied to vertical surfaces. Excellent for almost all surfaces. Ideal for porous substrates, as well as rough and acidic surfaces.
Catalog No. NC9613061 Supplier XPEDX Supplier No. 10910651
This item has a minimum qty of 41 per supplier requirements.
A general-purpose, no-run clear gel. Can be applied to vertical surfaces. Excellent for almost all surfaces. Ideal for porous substrates, as well as rough and acidic surfaces.