Learn More
Electron Microscopy Sciences Loctite Loctite 460 Adhesive
SDP
Catalog No. 501926789
Encompass_Preferred
Our Loctite 460 is a quick curing low viscosity glue that be used for mounting samples to any substrate (glass, metal and/or plastic). It is soluble in acetone. Comes in an applicator-tipped plastic bottle – 20g.
Catalog No. 50-192-6789 Supplier Electron Microscopy Sciences Supplier No. 1264608
Our Loctite 460 is a quick curing low viscosity glue that be used for mounting samples to any substrate (glass, metal and/or plastic). It is soluble in acetone. Comes in an applicator-tipped plastic bottle – 20g.