Learn More
RS Hughes Inc LOCTITE 498 SUPER GLUE,1OZ
SDP
Catalog No. NC9884114
Encompass_Preferred
Loctite Super Bonder 498 Cyanoacrylate Adhesive - Clear Liquid 1 oz Bottle - Shear Strength 3000 psi
Catalog No. NC9884114 Supplier RS Hughes Inc Supplier No. 07934049850
Loctite Super Bonder 498 Cyanoacrylate Adhesive - Clear Liquid 1 oz Bottle - Shear Strength 3000 psi