Learn More
Electron Microscopy Sciences Loctite Fixmaster Poxy Pak™ Fast Cure Epoxy
SDP
Supplier:  Electron Microscopy Sciences 1264609
Encompass_Preferred
This fast cure, general purpose epoxy bonds virtually any material. It will also fill or seal cracks and rebuild worn surfaces. Comes in a 1 oz. syringe with a built-in plunger for easy dispensing without equipment. Working time is approximately 4 minutes. Includes nozzle and mixing stick. This unique adhesive may be made electrically conductive with the addition of any conductive powder (Silver, Carbon, etc.)
Catalog No. 50-192-6790