Learn More
RS Hughes Inc Loctite Flash Cure 4310 Cyanoacrylate Adhesive, 1 oz
SDP
Catalog No. NC1678052
Encompass_Preferred
Loctite Flash Cure 4310 clear cyanoacrylate adhesive is compatible with metal, rubber and thermoplastic materials with a 24 hr cure time. Provides a 5 sec fixture time. Delivers great performance with a shear strength of 3190 psi, tensile strength of 7250 psi and peel strength of 17 pli.
Catalog No. NC1678052 Supplier RS Hughes Inc Supplier No. 07934000001
Loctite Flash Cure 4310 clear cyanoacrylate adhesive is compatible with metal, rubber and thermoplastic materials with a 24 hr cure time. Provides a 5 sec fixture time. Delivers great performance with a shear strength of 3190 psi, tensile strength of 7250 psi and peel strength of 17 pli.