Learn More
Electron Microscopy Sciences Loctite 4011™ Prism 20 GR
SDP
Catalog No. 50949057
Encompass_Preferred
For the assembly of difficult-to-bond materials, such as wood, paper, leather and fabric. Cure speed: 2 to 20 seconds depending on the material.
Catalog No. 50-949-057 Supplier Electron Microscopy Sciences Supplier No. 7257311
For the assembly of difficult-to-bond materials, such as wood, paper, leather and fabric. Cure speed: 2 to 20 seconds depending on the material.