Learn More
Electron Microscopy Sciences Loctite 454™ Prism Tube 3 GR
SDP
Catalog No. 50949016
Encompass_Preferred
For use with porous surfaces, clear, gap fills 0.010", surface insensitive Ethyl, gel type, tensile shear strength 3200 psi, temperature range-65 deg F to 180 deg F. Cure speed: fixture – 15 seconds, full - 24 hours
Catalog No. 50-949-016 Supplier Electron Microscopy Sciences Supplier No. 7257154
For use with porous surfaces, clear, gap fills 0.010", surface insensitive Ethyl, gel type, tensile shear strength 3200 psi, temperature range-65 deg F to 180 deg F. Cure speed: fixture – 15 seconds, full - 24 hours