Learn More
Electron Microscopy Sciences Loctite 4861™ Prism 20 GR
SDP
Catalog No. 50949056
Encompass_Preferred
For use with flexible surfaces, clear, gap fill 0.008", surface flexible Alkyl, viscosity 4,000 cP, tensile shear strength 2465 psi, temperature range-65 deg F to 212 deg F. Cure speed: fixture – 25 seconds, full – 24 hours.
Catalog No. 50-949-056 Supplier Electron Microscopy Sciences Supplier No. 7257261
For use with flexible surfaces, clear, gap fill 0.008", surface flexible Alkyl, viscosity 4,000 cP, tensile shear strength 2465 psi, temperature range-65 deg F to 212 deg F. Cure speed: fixture – 25 seconds, full – 24 hours.