Learn More
Uline Loctite Threadlocker 262, 50 mL
Catalog No. NC1685197
Encompass
One little drop prevents leakage. Seals metal threads. Permanent adhesive. High strength. 3/4" diameter and smaller fasteners. NSF-P1 registered.
Catalog No. NC1685197 Supplier Uline Supplier No. S15893
One little drop prevents leakage. Seals metal threads. Permanent adhesive. High strength. 3/4" diameter and smaller fasteners. NSF-P1 registered.