Learn More
Sigma Aldrich Fine Chemicals Biosciences Locust bean gum from Ceratonia siliqua seeds, 9000-40-2, MFCD00131257, 500 g
Catalog No. NC1151155
Encompass
Locust bean gum (LBG, galactomannan polysaccharide) from Ceratonia siliqua seeds may be used as a substrate to help identify, differentiate and characterize mannanase(s), such as the β-Mannanases (EC 3.2.1.78). LBG may be used in the development of drug delivery devices.
Catalog No. NC1151155 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G0753500G
Locust bean gum (LBG, galactomannan polysaccharide) from Ceratonia siliqua seeds may be used as a substrate to help identify, differentiate and characterize mannanase(s), such as the β-Mannanases (EC 3.2.1.78). LBG may be used in the development of drug delivery devices.