Learn More
Medchemexpress LLC Locust bean gum | 9000-40-2 | MFCD00131257 | >75.0% | ≈310,000 g/mol | (C6H10O5)n | 50 MG
SDP
Catalog No. 5000282234
Encompass_Preferred
Locust bean gum is a high-molecular-weight galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds. It acts as a thickening, stabilizing, and gelling agent for formulation and research applications and is supplied as an off-white to light-yellow solid intended for laboratory use.
  • Natural galactomannan polysaccharide with high molecular weight.
  • Functions as a thickener, stabilizer, and gelling agent.
  • High water retention and viscosity-enhancing properties.
  • Purity greater than 75% as typically specified for this grade.
  • Supplied as a solid suitable for formulation and analytical studies.
Catalog No. 50-002-82234 Supplier Medchemexpress LLC Supplier No. HYW25014650MG
Locust bean gum is a high-molecular-weight galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds. It acts as a thickening, stabilizing, and gelling agent for formulation and research applications and is supplied as an off-white to light-yellow solid intended for laboratory use.
  • Natural galactomannan polysaccharide with high molecular weight.
  • Functions as a thickener, stabilizer, and gelling agent.
  • High water retention and viscosity-enhancing properties.
  • Purity greater than 75% as typically specified for this grade.
  • Supplied as a solid suitable for formulation and analytical studies.