Learn More
Medchemexpress LLC Locust bean gum | 9000-40-2 | MFCD00131257 | >75.0% | ≈310 kDa | (C6H10O5)n | 250 MG
SDP
Catalog No. 5000282336
Encompass_Preferred
Locust bean gum is a natural galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds, provided as a powdered material for laboratory research use. It acts as a thickening, stabilizing, and gelling agent with good hydration properties and compatibility with other hydrocolloids.
  • Natural galactomannan polysaccharide with food-grade functionality.
  • Effective thickening and gelling at low concentrations.
  • High water-holding capacity and stability in cold conditions.
  • Compatible with other hydrocolloids to modify texture.
  • Useful as a stabilizer and binder in formulations.
Catalog No. 50-002-82336 Supplier Medchemexpress LLC Supplier No. HYW250146250MG
Locust bean gum is a natural galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds, provided as a powdered material for laboratory research use. It acts as a thickening, stabilizing, and gelling agent with good hydration properties and compatibility with other hydrocolloids.
  • Natural galactomannan polysaccharide with food-grade functionality.
  • Effective thickening and gelling at low concentrations.
  • High water-holding capacity and stability in cold conditions.
  • Compatible with other hydrocolloids to modify texture.
  • Useful as a stabilizer and binder in formulations.