Learn More
Medchemexpress LLC Locust bean gum | 9000-40-2 | MFCD00131257 | >75.0% | Approx. 310,000 g/mol | No single molecular formula - high-molecular-weight galactomannan polymer | 100 MG
SDP
Catalog No. 5000282153
Encompass_Preferred
Locust bean gum is a high-molecular-weight galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds and supplied for research use. It serves as a thickening, stabilizing, and gelling agent with strong water retention and high viscosity, commonly used in formulation studies, biochemical assays, and food technology research.
  • Natural galactomannan polysaccharide from carob seeds.
  • High viscosity (≈5000-6500 mPa·s) for robust thickening.
  • Polysaccharide purity >75%.
  • Forms stable gels at low temperatures.
  • Good water retention and suspension stabilization.
  • Storage: room temperature, protect from light; solutions: -20°C to -80°C for extended storage.
  • For research use only; not for patient or human therapeutic use.
Catalog No. 50-002-82153 Supplier Medchemexpress LLC Supplier No. HYW250146100MG
Locust bean gum is a high-molecular-weight galactomannan polysaccharide extracted from carob (Ceratonia siliqua) seeds and supplied for research use. It serves as a thickening, stabilizing, and gelling agent with strong water retention and high viscosity, commonly used in formulation studies, biochemical assays, and food technology research.
  • Natural galactomannan polysaccharide from carob seeds.
  • High viscosity (≈5000-6500 mPa·s) for robust thickening.
  • Polysaccharide purity >75%.
  • Forms stable gels at low temperatures.
  • Good water retention and suspension stabilization.
  • Storage: room temperature, protect from light; solutions: -20°C to -80°C for extended storage.
  • For research use only; not for patient or human therapeutic use.