Learn More
Medchemexpress LLC Locust bean gum from Ceratonia siliqua seeds | 9000-40-2 | MFCD00131257 | >75.0% | 856.8 g/mol | Not applicable | 500 MG
SDP
Catalog No. 5000281298
Encompass_Preferred
Locust bean gum is a natural galactomannan polysaccharide extracted from Ceratonia siliqua (carob) seeds. It is supplied as a powdered reagent used as a thickening, stabilizing, and gelling agent in food formulation, research, and laboratory applications (CAS 9000-40-2).
  • Thickens and stabilizes formulations.
  • Forms gels and improves texture in low-temperature systems.
  • High water retention for moisture control.
  • Compatible with other hydrocolloids such as xanthan and guar.
  • Typical polysaccharide content greater than 75%.
Catalog No. 50-002-81298 Supplier Medchemexpress LLC Supplier No. HYW250146500MG
Locust bean gum is a natural galactomannan polysaccharide extracted from Ceratonia siliqua (carob) seeds. It is supplied as a powdered reagent used as a thickening, stabilizing, and gelling agent in food formulation, research, and laboratory applications (CAS 9000-40-2).
  • Thickens and stabilizes formulations.
  • Forms gels and improves texture in low-temperature systems.
  • High water retention for moisture control.
  • Compatible with other hydrocolloids such as xanthan and guar.
  • Typical polysaccharide content greater than 75%.