Learn More
Echelon Biosciences Research Labs Locustapyrokinin, L. Migratoria - 5 mg, CAS#: 132293-87-9
SDP
Supplier:  Echelon Biosciences Research Labs 361255MG
Encompass_Preferred
Locustapyrokinin, L. Migratoria - 5mg , Glp-Asp-Ser-Gly-Asp-Gly-Trp-Pro-Gln-Gln-Pro-Phe-Val-Pro-Arg-Leu-NH2 This item is non returnable
Catalog No. 50-190-8430