Learn More
Medchemexpress LLC HY-100240 5mg Medchemexpress, Lodelaben CAS:111149-90-7 Purity:>98%
SDP
Catalog No. 502080580
Encompass_Preferred
Medchemexpress, HY-100240 5mg Lodelaben CAS:111149-90-7 Lodelaben is a human neutrophil elastase inhibitor with an IC50 and Ki of 0.5 and 1.5 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0580 Supplier Medchemexpress LLC Supplier No. HY1002405MG
Medchemexpress, HY-100240 5mg Lodelaben CAS:111149-90-7 Lodelaben is a human neutrophil elastase inhibitor with an IC50 and Ki of 0.5 and 1.5 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.