Learn More
Medchemexpress LLC HY-123210 5mg Medchemexpress, Lodenafil CAS:139755-85-4 Purity:>98%
SDP
Catalog No. 501932577
Encompass_Preferred
Medchemexpress, HY-123210 5mg Lodenafil CAS:139755-85-4 Lodenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor for the treatment of erectile dysfunction (ED) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2577 Supplier Medchemexpress LLC Supplier No. HY1232105MG
Medchemexpress, HY-123210 5mg Lodenafil CAS:139755-85-4 Lodenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor for the treatment of erectile dysfunction (ED) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.