Learn More
Medchemexpress LLC HY-108045 1mg Medchemexpress, Lodenafil carbonate CAS:398507-55-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1080451MG
Encompass_Preferred
Medchemexpress, HY-108045 1mg Lodenafil carbonate CAS:398507-55-6 Lodenafil carbonate, a dimer that acts as a prodrug delivering Lodenafil in vivo, is an orally active phosphodiesterase type 5 (PDE5) inhibitor for the treatment of erectile dysfunction (ED) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2309