Learn More
Selleck Chemical LLC Lodoxamide E4875-1G
SDP
Catalog No. 5000043308
Encompass_Preferred
Lodoxamide (U-42585E free acid) is a potent agonist of GPR35 with an EC50 of 1 nM It also inhibits eosinophil peroxidase release after IgA activation and reduces eosinophil cationic protein and eosinophil-derived neurotoxin suggesting direct effects on eosinophils It is a mast cell stabilizer used to treat asthma allergic conjunctivitis and superior limbic keratoconjunctivitis
Catalog No. 50-000-43308 Supplier Selleck Chemical LLC Supplier No. E48751G
Lodoxamide (U-42585E free acid) is a potent agonist of GPR35 with an EC50 of 1 nM It also inhibits eosinophil peroxidase release after IgA activation and reduces eosinophil cationic protein and eosinophil-derived neurotoxin suggesting direct effects on eosinophils It is a mast cell stabilizer used to treat asthma allergic conjunctivitis and superior limbic keratoconjunctivitis