Learn More
TARGETMOL CHEMICALS INC LODOXAMIDE TROMETHAMINE 5MG
SDP
Catalog No. 502581203
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lodoxamide tromethamine is a mast cell stabilizer with anti-allergic properties and can be used in studies about the treatment of asthma and allergic conjunctivitis. purity: 99%
Catalog No. 50-258-1203 Supplier TARGETMOL CHEMICALS INC Supplier No. T15772
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lodoxamide tromethamine is a mast cell stabilizer with anti-allergic properties and can be used in studies about the treatment of asthma and allergic conjunctivitis. purity: 99%