Learn More
Medchemexpress LLC Lodoxamide tromethamine | 63610-09-3 | 99.8% | 553.90 | 100 MG
SDP
Catalog No. 5000393952
Encompass_Preferred
Lodoxamide tromethamine is a medication used for the treatment and prophylaxis of mast cell-mediated allergic diseases. It demonstrates cromolyn-like activity and can inhibit histamine release and calcium influx in mast cells. It has shown to reduce allergic responses in vivo and can decrease lung reperfusion injury. Patients treated with this compound have reported improvements in breathing, cough, sputum production, and sleep.
  • Treatment and prophylaxis of mast cell-mediated allergic disease.
  • Inhibits compound 48/80-induced histamine release and calcium influx in mast cells.
  • Demonstrates cromolyn-like activity.
  • Reduces increased respiratory frequency and decreased tidal volume induced by antigen challenge.
  • Decreases lung reperfusion injury.
  • Improves daytime breathing difficulty, cough, sputum production, and sleep.
Catalog No. 50-003-93952 Supplier Medchemexpress LLC Supplier No. HY16289100MG
Lodoxamide tromethamine is a medication used for the treatment and prophylaxis of mast cell-mediated allergic diseases. It demonstrates cromolyn-like activity and can inhibit histamine release and calcium influx in mast cells. It has shown to reduce allergic responses in vivo and can decrease lung reperfusion injury. Patients treated with this compound have reported improvements in breathing, cough, sputum production, and sleep.
  • Treatment and prophylaxis of mast cell-mediated allergic disease.
  • Inhibits compound 48/80-induced histamine release and calcium influx in mast cells.
  • Demonstrates cromolyn-like activity.
  • Reduces increased respiratory frequency and decreased tidal volume induced by antigen challenge.
  • Decreases lung reperfusion injury.
  • Improves daytime breathing difficulty, cough, sputum production, and sleep.