Learn More
Medchemexpress LLC Lodoxamide tromethamine | 63610-09-3 | 99.8% | 553.90 | 50 MG
SDP
Catalog No. 5000394638
Encompass_Preferred
Lodoxamide tromethamine (U-42585E) is a medication for the treatment and prophylaxis of mast cell-mediated allergic disease. It inhibits histamine release and ionophore-induced 45Ca influx in purified rat peritoneal mast cells. In vivo, it demonstrates cromolyn-like activity and significantly inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge. It also helps decrease lung reperfusion injury and improves breathing difficulties, cough, sputum production, and sleep.
  • Treats and prevents mast cell-mediated allergic disease
  • Inhibits histamine release
  • Inhibits ionophore-induced 45Ca influx
  • Demonstrates cromolyn-like activity
  • Inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge
  • Decreases lung reperfusion injury
  • Improves daytime breathing difficulty, cough, sputum production, and sleep
Catalog No. 50-003-94638 Supplier Medchemexpress LLC Supplier No. HY1628950MG
Lodoxamide tromethamine (U-42585E) is a medication for the treatment and prophylaxis of mast cell-mediated allergic disease. It inhibits histamine release and ionophore-induced 45Ca influx in purified rat peritoneal mast cells. In vivo, it demonstrates cromolyn-like activity and significantly inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge. It also helps decrease lung reperfusion injury and improves breathing difficulties, cough, sputum production, and sleep.
  • Treats and prevents mast cell-mediated allergic disease
  • Inhibits histamine release
  • Inhibits ionophore-induced 45Ca influx
  • Demonstrates cromolyn-like activity
  • Inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge
  • Decreases lung reperfusion injury
  • Improves daytime breathing difficulty, cough, sputum production, and sleep