Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lodoxamide tromethamine | 63610-09-3 | MFCD04112765 | 99.8% | 553.9 g/mol | C19H28ClN5O12 | 10 MG
SDP

Catalog No. 5000288591
Encompass_Preferred

Lodoxamide tromethamine is a mast-cell stabilizer used in research to prevent and treat mast cell-mediated allergic responses. It is supplied as a solid and is also available as ready-to-use DMSO solutions for in vitro studies, and is intended for research use only.

  • Acts as a mast-cell stabilizer for prophylaxis of mast cell-mediated allergic disease.
  • Supplied as a solid with white to light yellow appearance.
  • High purity (about 99.8%).
  • Molecular weight 553.9 g/mol; chemical formula C19H28ClN5O12; CAS 63610-09-3.
  • Available as 10 mM solutions in DMSO (1 mL) and multiple solid pack sizes for experimental flexibility.
  • Storage: keep sealed and away from moisture; in solvent store at -80°C for long term or -20°C for short term.

Catalog No. 50-002-88591 Supplier Medchemexpress LLC Supplier No. HY1628910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lodoxamide tromethamine is a mast-cell stabilizer used in research to prevent and treat mast cell-mediated allergic responses. It is supplied as a solid and is also available as ready-to-use DMSO solutions for in vitro studies, and is intended for research use only.

  • Acts as a mast-cell stabilizer for prophylaxis of mast cell-mediated allergic disease.
  • Supplied as a solid with white to light yellow appearance.
  • High purity (about 99.8%).
  • Molecular weight 553.9 g/mol; chemical formula C19H28ClN5O12; CAS 63610-09-3.
  • Available as 10 mM solutions in DMSO (1 mL) and multiple solid pack sizes for experimental flexibility.
  • Storage: keep sealed and away from moisture; in solvent store at -80°C for long term or -20°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.