Learn More
Medchemexpress LLC Lodoxamide (U-42585E free acid) | 53882-12-5 | 98.1% | C11H6ClN3O6 | 200 MG
SDP
Catalog No. 5000353715
Encompass_Preferred
Lodoxamide (U-42585E free acid) is an antiallergic compound that acts as a mast-cell stabilizer, used for the treatment of asthma and allergic conjunctivitis.
  • Inhibits histamine release and calcium influx in mast cells.
  • Reduces chemotactic response of eosinophils to fMLP and IL-5.
  • Inhibits eosinophil peroxidase release after IgA-dependent activation.
  • Demonstrates cromolyn-like activity in mast cell assays and antigen-sensitized rhesus monkeys.
  • Inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge.
  • Reduces lung reperfusion injury in organ preservation solutions.
  • Improves daytime breathing difficulty, cough, sputum, and sleep in patients.
Catalog No. 50-003-53715 Supplier Medchemexpress LLC Supplier No. HY14270200MG
Lodoxamide (U-42585E free acid) is an antiallergic compound that acts as a mast-cell stabilizer, used for the treatment of asthma and allergic conjunctivitis.
  • Inhibits histamine release and calcium influx in mast cells.
  • Reduces chemotactic response of eosinophils to fMLP and IL-5.
  • Inhibits eosinophil peroxidase release after IgA-dependent activation.
  • Demonstrates cromolyn-like activity in mast cell assays and antigen-sensitized rhesus monkeys.
  • Inhibits increased respiratory frequency and decreased tidal volume induced by antigen challenge.
  • Reduces lung reperfusion injury in organ preservation solutions.
  • Improves daytime breathing difficulty, cough, sputum, and sleep in patients.